Learn More
Invitrogen™ ViewRNA™ Cell Plus Fixation/Permeabilization Buffer Set
Catalog No. 0019001
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
00-190-01 1 Kit
1 options
Catalog No. 00-190-01 Supplier Invitrogen™ Supplier No. 0019001
Add to Cart
Add to Cart
The ViewRNA™ Cell Plus Fixation/Permeabilization Buffer Set is a component of the ViewRNA™ Cell Plus Assay kit (Cat. No. 88-19000-99).
The ViewRNA™ Cell Plus Fixation/Permeabilization Buffer Set is a component of the ViewRNA™ Cell Plus Assay kit (Cat. No. 88-19000-99). These buffers can be used to evaluate antibody performance following the fixation and permeabilization steps of the assay. Note that additional changes to antibody performance may be observed following the hybridization steps of the ViewRNA™ Cell Plus Assay.
Please refer to the ViewRNA™ Cell Plus Assay protocol on the product data sheet for instructions for use.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage • ViewRNA™ Cell Plus Fixation/Permeabilization Component A (Cat. No. 00-19500-23)
• ViewRNA™ Cell Plus Fixation/Permeabilization Component B (Cat. No, 00-19900-23)
• ViewRNA™ Cell Plus Blocking/Antibody Diluent (Cat. No. 00-19600-54)
Store all at 2°C to 8°C.
Product Type Fixn/Perm Buffer Set
Quantity 1 Kit
Solution Type Fixation/Permeabilization Buffer
For Research Use Only. Not for use in diagnostic procedures.