Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ViewRNA™ microRNA, 1.44 mL

Catalog No. CVM02
Encompass_Preferred
Change view
Click to view available options
Quantity:
1.44 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CVM02 1.44 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CVM02 Supplier Invitrogen™ Supplier No. CVM02
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This is a customizable microRNA probe set for the ViewRNA ISH Cell, ViewRNA ISH Tissue, and ViewRNA Cell Plus assays.

This is a customizable microRNA probe set for the ViewRNA ISH Cell, ViewRNA ISH Tissue, and ViewRNA Cell Plus assays.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage ViewRNA ISH microRNA probe set, 1.44 mL

Store at -20°C
Quantity 1.44 mL
Product Line ViewRNA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.