Learn More
Invitrogen™ ViewRNA™ microRNA, 440 μL
Catalog No. CVM01
Encompass_Preferred
Change view
Click to view available options
Quantity:
440 μl
Catalog No. Quantity
CVM01 440 μl
1 options
Catalog No. CVM01 Supplier Invitrogen™ Supplier No. CVM01
Add to Cart
Add to Cart
This is a customizable microRNA probe set for the ViewRNA ISH Cell, ViewRNA ISH Tissue, and ViewRNA Cell Plus assays.
This is a customizable microRNA probe set for the ViewRNA ISH Cell, ViewRNA ISH Tissue, and ViewRNA Cell Plus assays.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage ViewRNA ISH microRNA probe set, 440 μL
Store at -20°C
Quantity 440 μl
Product Line ViewRNA