Learn More
Invitrogen™ ViewRNA™ Temperature Validation Kit
Catalog No. QV0523
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
QV0523 1 Kit
1 options
Catalog No. QV0523 Supplier Invitrogen™ Supplier No. QV0523
  • $532.00
    $483.65 / Each of 1
    Save $48.35
    9% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
The ViewRNA Temperature Validation Kit uses a calibrated digital thermometer to assess temperature accuracy.
The ViewRNA Temperature Validation Kit uses a calibrated digital thermometer to assess temperature accuracy. This kit may be used for any ViewRNA Assay to evaluate the temperature of incubators or ThermoBrite systems.
TRUSTED_SUSTAINABILITY
Specifications
Type Temperature Validation Kit
Quantity 1 Kit
For Research Use Only. Not for use in diagnostic procedures.