Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ViewRNA Tissue Assay Fluorescence Kits

Catalog No. QVT4694B
Encompass_Preferred
Change view
Click to view available options
Product Type:
Tissue Alexa Fluor 488 Module
Tissue Alexa Fluor 546 Module
Tissue Alexa Fluor 594 Module
Tissue Alexa Fluor 750 Module
Tissue Fluor. 1 Plex Assay
Tissue Fluor. 4 Plex Assay Kit
Color:
Deep Red (Cy5)
Green (FITC)
Green, Orange, Deep Red, NIR
Green, Red, Deep Red, NIR
Near Infrared (Cy7)
Orange (TRITC)
Red (Texas Red)
Quantity:
24 Slides
96 Slides
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color Quantity
QVT4694B Tissue Alexa Fluor 594 Module Red (Texas Red) 96 Slides
QVT0700 Tissue Fluor. 4 Plex Assay Kit Green, Orange, Deep Red, NIR 24 Slides
QVT4700 Tissue Fluor. 4 Plex Assay Kit Green, Orange, Deep Red, NIR 96 Slides
QVT0800 Tissue Fluor. 4 Plex Assay Kit Green, Red, Deep Red, NIR 24 Slides
QVT4800 Tissue Fluor. 4 Plex Assay Kit Green, Red, Deep Red, NIR 96 Slides
QVT0600C Tissue Fluor. 1 Plex Assay Deep Red (Cy5) 24 Slides
QVT4600C Tissue Fluor. 1 Plex Assay Deep Red (Cy5) 96 Slides
QVT0688B Tissue Alexa Fluor 488 Module Green (FITC) 24 Slides
QVT4688B Tissue Alexa Fluor 488 Module Green (FITC) 96 Slides
QVT0646B Tissue Alexa Fluor 546 Module Orange (TRITC) 24 Slides
QVT4646B Tissue Alexa Fluor 546 Module Orange (TRITC) 96 Slides
QVT0694B Tissue Alexa Fluor 594 Module Red (Texas Red) 24 Slides
QVT0640B Tissue Alexa Fluor 750 Module Near Infrared (Cy7) 24 Slides
QVT4640B Tissue Alexa Fluor 750 Module Near Infrared (Cy7) 96 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Kit: Danger. Contains Formamide and Subtilisin. Suspected of causing cancer. May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Contains Lithium Dodecyl Sulphate. Causes skin irritation. Causes serious eye irritation. Read SDS.
Catalog No. QVT4694B Supplier Thermo Scientific™ Supplier No. QVT4694B
Only null left
Add to Cart
Add to Cart
Kit: Danger. Contains Formamide and Subtilisin. Suspected of causing cancer. May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Contains Lithium Dodecyl Sulphate. Causes skin irritation. Causes serious eye irritation. Read SDS.

Requires

When stored as directed on product label, product is stable for at least 6 months after receipt. Lot specific expiration date can be found on certificate of analysis (COA).

ViewRNA Tissue assays are RNA in situ hybridization (ISH) assays for the reliable detection of up to four RNA targets within tissue sections and cultured cells. Visualize any RNA in any tissue/cell with single-copy sensitivity.

ViewRNA Tissue Assay Fluorescence kits enable an in situ hybridization (ISH) method for the simultaneous detection of one to four RNA targets (mRNA, non-coding RNA, or microRNA) at single-copy sensitivity. These kits combine bright Alexa Fluor 488, Alexa Fluor 594, Alexa Fluor 546, Alexa Fluor 647, and Alexa Fluor 750 dyes that are visualized using fluorescence microscopes or high-content imagers. ViewRNA Tissue assay uses proprietary chemistry for the target-specific probe sets (RNA FISH probes) and branch DNA (bDNA) signal amplification for detection of specific signal.

With ViewRNA Tissue assays, you can achieve:

  • Robust, single RNA molecule detection in FFPE tissue sections, cryo-preserved sections, and cultured cells
  • Accuracy and specificity inherent to bDNA amplification technologies
  • Simultaneous detection of up to four RNA targets, including mRNA, noncoding RNA, and microRNA

Note: A ViewRNA probe set is required and can be purchased separately

ViewRNA Tissue Assay Fluorescence kits, when combined with a ViewRNA Tissue probe set (purchased separately), enables the detection of a single RNA target in tissues/cells using Alexa Fluor dyes. ViewRNA offers 6500 catalog probe sets. Any more probe sets can be designed at no extra cost.

To meet various experimental needs, the ViewRNA Tissue assay is provided in various kits and modules.

ViewRNA Tissue Fluorescence 4-Plex Assay Kit (green, orange, deep red, and near infrared)

With this kit four RNA targets can be detected using ViewRNA probe sets type 1, 4, 6, and 10 (ViewRNA probe sets are sold separately). This kit utilizes Alexa Fluor 546 for Type 1 probes, Alexa Fluor 488 for Type 4 probes, Alexa Fluor 647 for Type 6 probes, and Alexa Fluor 750 for Type 10 probes. RNA visualization requires FITC/GFP, TRITC/RFP, CY5/Deep Red, and Cy7/NIR microscope filter sets. An additional DAPI filter set is needed for nucleus counterstain with DAPI.

ViewRNA Tissue Fluorescence 4-Plex Assay Kit (green, red, deep red, and near infrared)

With this kit four RNA targets can be detected using ViewRNA probe sets type 1, 4, 6, and 10 (ViewRNA probe sets are sold separately). This kit utilizes Alexa Fluor 594 for Type 1 probes, Alexa Fluor 488 for Type 4 probes, Alexa Fluor 647 for Type 6 probes, and Alexa Fluor 750 for Type 10 probes. RNA visualization requires FITC/GFP, Texas Red, CY5/Deep Red, and Cy7/NIR microscope filter sets. An additional DAPI filter set is needed for nucleus counterstain with DAPI.

ViewRNA Tissue Fluorescence 1-Plex Assay Core Kit, with Alexa Fluor 647 (Type 6) label probe

With this kit, one RNA target can be detected using a Type 6 ViewRNA probe set (ViewRNA probe sets are sold separately). Up to four RNA targets can be detected when combined with ViewRNA Tissue modules (sold separately). RNA visualization requires a Cy5/Deep Red microscope filter set. An additional DAPI filter set is needed for nucleus counterstain with DAPI.

ViewRNA Tissue Alexa Fluor 488 (Type 4) Module

When combined with the ViewRNA Tissue Fluorescence 1-Plex Assay Core Kit (Cat. No. QVT0600) this module can detect an additional RNA target utilizing a Type 4 ViewRNA probe set. This module can be combined with other modules for additional RNA target detection. RNA visualization requires a FITC/GFP filter set.

ViewRNA Tissue Alexa Fluor 546 (Type 1) Module

When combined with the ViewRNA Tissue Fluorescence 1-Plex Assay Core Kit (Cat. No. QVT0600) this module can detect an additional RNA target utilizing a Type 1 ViewRNA probe set. This module can be combined with other modules for additional RNA target detection. RNA visualization requires a TRITC/RFP filter set.

Note: The ViewRNA Tissue Alexa Fluor 546 (Type 1) Module cannot be combined with the ViewRNA Tissue Alexa Fluor 594 (Type 1) Module because both use a Type 1 probe set and will spectrally overlap in most fluorescence microscopes.

ViewRNA Tissue Alexa Fluor 594 (Type 1) Module

When combined with the ViewRNA Tissue Fluorescence 1-Plex Assay Core Kit (Cat. No. QVT0600) this module can detect an additional RNA target utilizing a Type 1 ViewRNA probe set. This module can be combined with other modules for additional RNA target detection. RNA visualization requires a Texas Red filter set.

Note: The ViewRNA Tissue Alexa Fluor 594 (Type 1) Module cannot be combined with the ViewRNA Tissue Alexa Fluor 546 (Type 1) Module because both use a Type 1 probe set and will spectrally overlap in most fluorescence microscopes.

ViewRNA Tissue Alexa Fluor 750 (Type 10) Module

When combined with ViewRNA Tissue Fluorescence 1-Plex Assay Core Kit (Cat. No. QVT0600) this module can detect an additional RNA target utilizing a Type 10 ViewRNA probe set. This module can be combined with other modules for additional RNA target detection. RNA visualization requires a CY7/NIR filter set.

TRUSTED_SUSTAINABILITY

Specifications

Color Red (Texas Red)
Detection Method Fluorescence
Format Kit
Label or Dye Alexa Fluor 594
Final Product Type DNA
Sample Type Tissue (FFPE), Tissue (CRYO), Cell Culture Cells
Labeling Target RNA (General)
Excitation/Emission 590/617 nm
For Use With (Application) RNA-FISH, In Situ Hybridization (ISH)
Labeling Method ViewRNA Type 1
Product Line ViewRNA
Product Type Tissue Alexa Fluor 594 Module
Quantity 96 Slides
Show More Show Less
How do ViewRNA assays compare to RNAScope assays?

ViewRNA and RNAScope technologies rely on the same signal amplification strategy - branched DNA amplification. Historically, both ViewRNA and RNAScope technologies originated from the same company, Panomics. The assays are expected to yield similar sensitivity and resolution, however each technology relies on its own set of proprietary reagents and probe set designs. Hence, the assays are not considered interchangeable or compatible. ViewRNA probe sets are not tested for RNAScope assaya and vice versa.

How is the signal amplified in ViewRNA assays?

The ViewRNA technology relies on branched DNA signal amplification strategy. Target probes complementary to the target transcript sequence are further hybridized with pre-amplifier, amplifier and label probes that consist of branched DNA, and form 'tree branches' that allow numerous label probes to attach. This approach allows higher signal amplification compared to traditional ISH techniques.

Where can I find general information about ViewRNA ISH Assays?

For general information about ViewRNA ISH Assays, please go to this page (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cellular-imaging/in-situ-hybridization-ish/rna-fish/viewrna-assays.html).

For ViewRNA assays, can I use the same set of wash solutions for all samples, including the positive and negative controls?

We do not recommend doing this. The negative control should be processed and washed separately from the rest of the samples. This is because the negative control does not contain any target probe sets and only the amplification reagents are added to it. If experimental samples are washed in the same beaker of wash solutions as the negative control, any unbound target probes that wash away can carry over to the negative control sample and cause unexpected positive signal (that will also appear to be very specific).

Is there a video on the ViewRNA tissue assay workflow that I can watch?

Yes, we do have a video that will help provide hands-on guidance on how to proceed with the protocol. Please find the link to the online protocol and video (https://www.thermofisher.com/us/en/home/references/protocols/cell-and-tissue-analysis/protocols/viewrna-tissue-fluorescence-protocol.html).

Do you have a list of antibodies that are compatible with ViewRNA assays?

Unfortunately, we do not have such a list as each antibody may perform differently on the varied sample types. We recommend testing each antibody in the context of the ViewRNA assay workflow to ascertain quality results. Assay conditions can be optimized by modifying the protease treatment time as well as optimizing heat pretreatment for antigen recovery.

Do you have a list of tissue types tested to work with ViewRNA Tissue Assay kits?

Tissue types that have been tested to work with ViewRNA Tissue Assay kits can be found listed on page 30 of the ViewRNA Tissue Fluorescence Assay User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0026643_ViewRNA_TissueFluorAssay_UG.pdf).

Tissue types that have been tested to work with ViewRNA Tissue Assay kits can be found listed on pages 29-30 of the ViewRNA Tissue Fluorescence Assay User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0026643_ViewRNA_TissueFluorAssay_UG.pdf) and on pages 48-49 of the ViewRNA Tissue Colorimetric Assay User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018633_viewRNA_ISH_UG.pdf).

Can I multiplex ViewRNA ISH tissue fluorescence assays with IHC or IF (antibody immunostaining)?

It is possible to combine RNA ISH on tissues with IF or IHC using immunofluorescence. It is essential to test the antibodies on the desired tissue type and under the conditions of the ViewRNA tissue assay workflow. Due to variable tissue processing conditions, not all antigens may remain intact. Note that antibodies added to the ViewRNA workflow should be RNAse-free or RNAse inhibitor-treated. Guidelines for combining RNA ISH and IHC can be found on page 31 of the ViewRNA Tissue Fluorescence Assay User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0026643_ViewRNA_TissueFluorAssay_UG.pdf).

Can I use the reagents from ViewRNA Tissue Assay Fluorescence kits and ViewRNA Colorimetric Tissue Assay kits interchangeably?

We do not recommend interchanging reagents between ViewRNA Tissue Assay Fluorescence kits and ViewRNA Colorimetric Tissue Assay kits. While some reagents, such as the Wash buffer components can be used interchangeably, other components such as the amplifier reagents are not formulated the same way.

What is supplied in the ViewRNA Fluorescence Tissue modules (Cat. No. QVT0688B, QVT0646B, QVT0694B, QVT0640B or QVT4688B, QVT4646B, QVT4694B, and QVT4640B)?

The ViewRNA Fluorescence Tissue modules provide the specific label probe, supplied as a standalone reagent:
- Cat. No. QVT0646B, QVT4646B: ViewRNA Tissue Alexa Fluor 546 (Type 1) Module
- Cat. No. QVT0694B, QVT4694B: ViewRNA Tissue Alexa Fluor 594 (Type 1) Module
- Cat. No. QVT0688B, QVT4688B: ViewRNA Tissue Alexa Fluor 488 (Type 4) Module
- Cat. No. QVT0640B, QVT4640B: ViewRNA Tissue Alexa Fluor 750 (Type 10) Module

Are all ViewRNA Tissue Assay kits capable of performing up to a 4-plex assay?

No, the ViewRNA Colorimetric Tissue Assay kits only allow up to a 2-plex assay. To perform a 2-plex colorimetric tissue assay, the ViewRNA Tissue Core Kit (Cat. No. QVT0400) can be used in combination with ViewRNA Modules purchased separately (Cat. No. QVT0410, QVT0420, QVT0430) or one of the ViewRNA Colorimetric Tissue 2-Plex kits can be used (Cat. No. QVT4433C, QVT0433C, QVT0412C, QVT4412C, QVT0422C, QVT4422C).

ViewRNA Tissue Assay Fluorescence kits allow up to a 4-plex assay with purchase of kit Cat. No. QVT0700, QVT4700, QVT0800, or QVT4800 or by using the ViewRNA Tissue Fluorescence 1 Plex Assay Core Kit (Cat. No. QVT0600C or QVT4600C) in combination with the pertinent Type 1 (Cat. No. QVT0646B, QVT4646B, QVT0694B, QVT4694B), Type 4 (Cat. No. QVT0688B, QVT4688B), or Type 10 (Cat. No. QVT0640B, QVT4640B) ViewRNA Tissue modules purchased separately.

What ViewRNA ISH assay kits do you offer for tissue samples?

ViewRNA ISH tissue assays are offered in two versions: colorimetric and fluorescence RNA ISH tissue assays.

- ViewRNA Tissue Assay Fluorescence Kits come with Alexa Fluor dye-conjugated label probes and enable the simultaneous detection of one to four RNA targets (mRNA, non-coding RNA, or microRNA) at single-copy sensitivity (Cat. No. QVT0700, QVT4700, QVT0800, QVT4800, QVT0600C, QVT4600C, QVT0688B, QVT4688B, QVT0646B, QVT4646B, QVT0694B, QVT4694B, QVT0640B, QVT4640B).

- ViewRNA Colorimetric Tissue Core Kit and Individual Modules rely on enzyme-mediated signal amplification, using AP- or HRP-conjugated label probes in conjunction with Fast Red, Fast Blue, or DAB substrates. These colorimetric assays enable detection of one or two mRNA targets within tissue sections at single-copy sensitivity (Cat. No. QVT0410, QVT0420, QVT0430, QVT0400, QVT0400A, QVT0400B).


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.