Learn More
Medchemexpress LLC HY-15399 10mg Medchemexpress, Vigabatrin CAS:68506-86-5 Purity:>98%
SDP
Catalog No. 502081717
Encompass_Preferred
Medchemexpress, HY-15399 10mg Vigabatrin CAS:68506-86-5 Vigabatrin (γ-Vinyl-GABA), an inhibitory neurotransmitter GABA vinyl-derivative, is an orally active and irreversible GABA transaminase inhibitor. Vigabatrin is an antiepileptic agent, which acts by increasing GABA levels in the brain by inhibiting the catabolism of GABA by GABA transaminase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1717 Supplier Medchemexpress LLC Supplier No. HY1539910MG
Medchemexpress, HY-15399 10mg Vigabatrin CAS:68506-86-5 Vigabatrin (γ-Vinyl-GABA), an inhibitory neurotransmitter GABA vinyl-derivative, is an orally active and irreversible GABA transaminase inhibitor. Vigabatrin is an antiepileptic agent, which acts by increasing GABA levels in the brain by inhibiting the catabolism of GABA by GABA transaminase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.