Learn More
Vikan™ Stainless Steel Handle, 39 in.
Catalog No. 17227155 Shop All Remco Products
Change view
Click to view available options
Color Code:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Code
17-227-155 Yellow
17-227-153 Red
17-227-151 Green
17-227-152 Blue
17-227-154 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-227-155 Supplier Vikan™ Supplier No. 29836
Add to Cart
Add to Cart
Handle is ideal for use in tight spaces where maneuverability is key.
  • Suitable for cleaning with the use of aggressive chemicals and where the handle is exposed to a heavy-handed treatment
  • The hygiene sleeve makes the handle more ergonomically friendly
  • Color-coded
Specifications
Color Code Yellow
Product Type Handle
Sterility Non-sterile
Quantity 1 Ea.
Description Thread type Euro Hygienic
Color-coded Yes
Material Stainless Steel