Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Telescopic Aluminum Handle, 62 to 113 in.

Catalog No. 17227144 Shop All Remco Products
Change view
Click to view available options
Color Code:
Black
Blue
Green
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Code
17-227-144 Black
17-227-141 Red
17-227-143 Yellow
17-227-139 Green
17-227-140 Blue
17-227-142 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-227-144 Supplier Vikan™ Supplier No. 29759
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Telescopic aluminum handle is a durable and safe option to use in hygiene sensitive areas.

  • For use with pad holder and deck scrubs
  • Suitable for reaching high walls and tanks
  • Cna be used with clean-in-place brushes for cleaning on top of overhead pipes
  • The handle extends to a maximum length of 115 in.
  • Uniquely suited for color-coding and HACCP programs

Specifications

Color Code Black
Product Type Handle
Sterility Non-sterile
Quantity 1 Ea.
Description Thread type Euro Hygienic
Certifications/Compliance FDA Compliant
Color-coded Yes
Material Aluminum, Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.