Learn More
Vikan™ Cleaning Pad Holder
Catalog No. 19830127 Shop All Remco Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-830-127 Red
17-227-468 Orange
17-227-467 Yellow
17-227-466 Green
19-830-126 Blue
19-830-128 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-830-127 Supplier Vikan™ Supplier No. 55004
Add to Cart
Add to Cart
Cleaning pad holder for cleaning underneath equipment and scrubbing stubborn dirt on walls, tanks, and floors.
  • Ideal for cleaning walls and tanks
  • Can be used for scrubbing stubborn dirt and cleaning underneath equipment
  • Stainless steel clip mechanism prevents the pad from falling off when heavy
  • Color-coded
  • European thread
Specifications
Sterility Non-sterile
Color Red
Diameter (English) 3.6 in.
Dimensions (L x W x H) 9 x 3.6 x 4.7 in.
Material Polypropylene, Stainless Steel, Nylon
Quantity 1 Ea.
Size Medium