Learn More
Description
- Supports any product and is virtually unbreakable
- Includes screws, wall anchors, and mounting instructions
- Provides a color-coded storage option for tools
- Proper storage of cleaning equipment
- Supports hygienic environments
- Helps to extend the usable life of the equipment
Specifications
Specifications
| Product Type | Wall Bracket |
| Color | Red |
| Material | Polypropylene |
| For Use With (Application) | Color-coded storage |
| Certifications/Compliance | FDA Compliant |
| Quantity | 1 |
| Dimensions (L x W x H) | 9.4 x 6.5 x 2.25 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.