Learn More
Vikan™ Ergonomic Shovel with Small Blade
Catalog No. 17227496 Shop All Remco Products
Change view
Click to view available options
Color Code:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Code
17-227-496 Red
17-227-498 Yellow
17-227-494 Green
17-227-495 Blue
17-227-497 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-227-496 Supplier Vikan™ Supplier No. 56114
Add to Cart
Add to Cart
Vikan™ Ergonomic Shovel with Small Blade is designed without any sharp corners or deep recessess.
  • Designed for use in direct contact with food
  • The handle is reinforced with an inner aluminum tube
  • It is stepped in two places to help prevent back injuries
  • The neck joint has been strengthened in order to withstand heavy-duty use
  • The long and small blade combination makes it suitable for shoveling smaller volumes and reaching into deep containers
  • Color-coded
Specifications
Product Type Ergonomic Shovel
Blade Material Polypropylene
Color Code Red
Color-coded Yes
Handle Material Polypropylene, Aluminum
Head Style Square
Length (English) 50 in.
Certifications/Compliance FDA Compliant
No. per Pack 1 Ea.
Weight (English) 3.39 lb.