Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Floor Brooms with Soft Bristles

Catalog No. 19830014 Shop All Remco Products
Change view
Click to view available options
Length (English):
24 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-830-014 24 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-830-014 Supplier Vikan™ Supplier No. 31994
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

With soft bristles for sweeping fine particles in dry areas.

  • Fully color-coded
  • Polypropylene block with polyester bristles
  • Autoclavable to 121°C

Specifications

Color Code Red
Product Type Soft Bristles
Length (English) 24 in.
For Use With (Application) Sweeping Fine Particles in Dry Areas
Color-coded Yes
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.