Learn More
Vikan™ Handheld Cleaning Pad Holder
Catalog No. 17227471 Shop All Remco Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-471 Red
17-227-473 Yellow
17-227-469 Green
17-227-470 Blue
17-227-472 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-227-471 Supplier Vikan™ Supplier No. 55104
Add to Cart
Add to Cart
Cleaning pad holder can be used with one of three types of pads to clean stainless steel, floors, and walls.
  • Tool for cleaning walls and tanks
  • Ideal for scrubbing stubborn dirt and cleaning underneath equipment
  • The clip mechanism is stainless steel, which prevents the pad from falling off once heavy with water
  • Color-coded
Specifications
Sterility Non-sterile
Color Red
Diameter (English) 3.8 in.
Dimensions (L x W x H) 9.1 x 3.8 x 3 in.
Material Polypropylene, Stainless Steel, Nylon
Quantity 1 Ea.
Size Medium