Learn More
MSC Vikan Push Broom Head, Heavy-Duty, 16.5 in, Soft/Stiff Bristles, White
Catalog No. 502836316
Encompass
This heavy-duty push broom head is engineered for efficient cleaning in demanding environments, particularly in food and beverage production areas. Featuring a durable polypropylene block and a combination of soft and stiff polyester bristles, it effectively manages both fine particles and larger debris. Its 16.5 in length and European threaded connection ensure thorough cleaning performance.
  • Heavy-duty construction
  • Soft and stiff polyester bristles for fine and large debris
  • Durable polypropylene block
  • Flared bristles reach into corners and wall junctions
  • Angle-cut bristles prevent skipping during long pushes
  • Heavier build maintains better floor contact for efficient cleaning
  • Color-coded to reduce cross-contamination
  • Compatible with European threaded handles (sold separately)
  • Certified for EU 10/2011 on Food Contact
Catalog No. 50-283-6316 Supplier MSC Supplier No. 45845369
This heavy-duty push broom head is engineered for efficient cleaning in demanding environments, particularly in food and beverage production areas. Featuring a durable polypropylene block and a combination of soft and stiff polyester bristles, it effectively manages both fine particles and larger debris. Its 16.5 in length and European threaded connection ensure thorough cleaning performance.
  • Heavy-duty construction
  • Soft and stiff polyester bristles for fine and large debris
  • Durable polypropylene block
  • Flared bristles reach into corners and wall junctions
  • Angle-cut bristles prevent skipping during long pushes
  • Heavier build maintains better floor contact for efficient cleaning
  • Color-coded to reduce cross-contamination
  • Compatible with European threaded handles (sold separately)
  • Certified for EU 10/2011 on Food Contact