Learn More
MSC Vikan Push Broom, 16.5 inch, Soft/Stiff Polyester Bristle, Polypropylene Block
Supplier: MSC 45845211
The Vikan Heavy-Duty Push Broom is designed for efficient cleaning in demanding environments. It features a heavier head for better floor contact, angle-cut bristles to prevent skipping, and flared bristles that reach into corners. With densely packed stiff and soft polyester bristles, it effectively sweeps both fine and larger debris.
- Designed based on feedback from US food and beverage manufacturers.
- Heavier broom head for easier contact with the floor during sweeping.
- Angle cut bristles prevent skipping or hopping during long pushes across production flooring.
- Flared bristles reach into corners and wall junctions for easier and more thorough cleaning.
- Densely packed stiff and soft bristles sweep both fine and larger debris.
- Chemical resistant for diverse cleaning environments.
- Multi-purpose application for varied cleaning needs.