Learn More
Vikan™ Nylon Flex Rod Handle
Catalog No. 19830229 Shop All Remco Products
Change view
Click to view available options
Color:
White
Catalog No. Color
19-830-229 White
1 options
Catalog No. 19-830-229 Supplier Vikan™ Supplier No. 53525
Add to Cart
Add to Cart
Nylon flex rod handle is used with a tube brush for cleaning around sensitive areas such as glass.
  • Used with a tube brush for cleaning around sensitive areas such as glass
  • Flexibility allows it to follow curved pipes
  • The handle combines with tube cleaners, part numbers 5354, 5356, 5365, 5368, and 5391 to deliver the ultimate flexibility in cleaning
  • Color-coded
Specifications
Color Code White
Product Type Handle
Color White
Sterility Non-sterile
Quantity 1 Ea.
Color-coded Yes
Material Polyamide, Polypropylene