Learn More
Vikan™ Stiff Paddle Scraper
Catalog No. 17227742 Shop All Remco Products
Catalog No. 17-227-742 Supplier Vikan™ Supplier No. 70114
Only null left
Add to Cart
Add to Cart
Stiff paddle scraper is used to remove remaining food stuff from containers prior to the cleaning procedure.
  • Intended for scraping substances from the sides of containers
  • Ideal tool for prepping prior to the cleaning procedure
  • Euro-thread connection allows for pairing with any length or type of Vikan™ handle
  • Can be used on everything from deep containers to simple mixing bowls
  • Rigid construction is useful when firm pressure needs to be applied into deep containers
  • Color-coded
Specifications
Product Type Scraper
Color-coded Yes
Color Code Red
Certifications/Compliance FDA Compliant
Color Red
Material Nylon
Size Medium
Description Thread type Euro Hygienic
Dimensions (L x W) 8.75 x 4.3 in.
Quantity 1 Ea.
Includes Thread type Euro Hygienic
Weight (English) 0.32 lb.
No. per Pack 1 Ea.