This table and floor scraper is designed for efficient removal of stubborn debris from floors and other surfaces. It features a robust polypropylene body and a high-quality AISI 304 stainless steel blade, ensuring durability and effective cleaning. The product is compliant with stringent EU and US FDA food contact regulations, making it suitable for use in environments requiring high hygiene standards.
- Effectively removes stubborn debris from surfaces.
- Constructed from durable polypropylene.
- Features a high-quality AISI 304 stainless steel blade.
- Complies with EU food contact regulations.
- Complies with US FDA food contact regulations.
- Suitable for contact with aqueous, acidic, alcoholic, fatty, and dry food types.
- Withstands heating up to 70 °C for 2 hours, or 100 °C for 15 minutes.
- Designed for easy cleaning and disinfection.