Learn More
Vikan™ Telescopic Fiberglass Handle
Catalog No. 17227145 Shop All Remco Products
Change view
Click to view available options
Color:
Gray
Catalog No. Color
17-227-145 Gray
1 options
Catalog No. 17-227-145 Supplier Vikan™ Supplier No. 2977
Add to Cart
Add to Cart
Telescopic fiberglass handle features an easy extending action, which makes it an ideal choice for cleaning walls and tanks where tool length flexibility is required.
  • Can be used with all brooms, squeegees, and scrapers
  • The extra length makes it an ideal handle for cleaning walls and tanks, as well as removing condensation from pipes
  • The fiberglass material makes the handle durable and long-lasting
  • Color-coded
Specifications
Color Code Gray
Product Type Telescopic Handle
Color Gray
Sterility Non-sterile
Quantity 1 Ea.
Description Thread type Euro Hygienic
Color-coded Yes
Material Fiberglass, Polypropylene