Learn More
Vikan™ Waterfed Telescopic Handle with Quick Disconnect Fitting
Catalog No. 17227138 Shop All Remco Products
Change view
Click to view available options
Color:
White
Catalog No. Color
17-227-138 White
1 options
Catalog No. 17-227-138 Supplier Vikan™ Supplier No. 2973Q5
Add to Cart
Add to Cart
Vikan™ Waterfed Telescopic Handle with Quick Disconnect Fitting allows water to travel through it for use in conjunction with a condensation squeegee or waterfed brush system.
  • The male quick disconnect fitting pairs with a female quick connect adapter, 0703US, which allows it to be conveniently used with a water hose for a continuous stream of water
  • Color-coded
Specifications
Product Type Waterfed Telescopic Handle
Color-coded Yes
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color White
Description Thread type Euro Hygienic, Quick Connect
Includes Thread type Euro Hygienic, Quick Connect
Length (English) 62.9 to 113.9 in.
Material Polypropylene
Quantity 1 Ea.
Width (English) 1.25 in.