Learn More
MSC VIKING Hi-Vis Rain Bib Pants, 5XL, 1 ea
Catalog No. 5000677374
Encompass
These hi-visibility rain bib pants are constructed from polyester and polyurethane, offering protection and enhanced visibility in wet working conditions. They are designed for outdoor environments requiring high visibility and water resistance.
  • Bib pant style for extended coverage
  • Polyester and polyurethane construction
  • Men's 5XL size with 34 1/2 in. inseam and 51 in. maximum waist size
  • Green/Yellow high-visibility color
  • ANSI Class E compliant for safety
  • Includes polyester lining for comfort
  • Hook-and-loop ankle closure for secure fit
  • Fabric weight of 6.75 oz.
  • Pull-on fly and elastic waist closure
  • Meets ANSI/ISEA 107-2015 Class E and CSA Z96-15 Class 2, Level 2 standards
Catalog No. 50-006-77374 Supplier MSC Supplier No. 42331488
These hi-visibility rain bib pants are constructed from polyester and polyurethane, offering protection and enhanced visibility in wet working conditions. They are designed for outdoor environments requiring high visibility and water resistance.
  • Bib pant style for extended coverage
  • Polyester and polyurethane construction
  • Men's 5XL size with 34 1/2 in. inseam and 51 in. maximum waist size
  • Green/Yellow high-visibility color
  • ANSI Class E compliant for safety
  • Includes polyester lining for comfort
  • Hook-and-loop ankle closure for secure fit
  • Fabric weight of 6.75 oz.
  • Pull-on fly and elastic waist closure
  • Meets ANSI/ISEA 107-2015 Class E and CSA Z96-15 Class 2, Level 2 standards