Learn More
MSC Viking Hi-Vis Rain Bib Pants, ANSI E, L, 1 ea
Catalog No. 5000783018
Encompass
These Hi-Visibility Rain Bib Pants are constructed from 300D Trilobal Rip-Stop Polyester, designed to provide durable and waterproof protection in demanding outdoor environments. They feature high-visibility properties to ensure safety and are suitable for various professional applications.
  • Material: 300D Trilobal Rip-Stop Polyester
  • High-visibility design
  • Resists abrasion, puncture, rip, and snag
  • Waterproof and windproof
  • Double stitched, heat sealed, and taped seams
  • Includes polyester lining
  • Drawstring waist closure with hook-and-loop fly access
  • Ankle type: Hook-and-loop
  • ANSI/ISEA 107-2015 Class E and CSA Z96-15 Class 2, Level 2 compliant
Catalog No. 50-007-83018 Supplier MSC Supplier No. 77801033
These Hi-Visibility Rain Bib Pants are constructed from 300D Trilobal Rip-Stop Polyester, designed to provide durable and waterproof protection in demanding outdoor environments. They feature high-visibility properties to ensure safety and are suitable for various professional applications.
  • Material: 300D Trilobal Rip-Stop Polyester
  • High-visibility design
  • Resists abrasion, puncture, rip, and snag
  • Waterproof and windproof
  • Double stitched, heat sealed, and taped seams
  • Includes polyester lining
  • Drawstring waist closure with hook-and-loop fly access
  • Ankle type: Hook-and-loop
  • ANSI/ISEA 107-2015 Class E and CSA Z96-15 Class 2, Level 2 compliant