This foul weather jacket provides thermal protection and FR compliance for industrial sites. Constructed from heavy-duty FR-treated 300 denier Trilobal ripstop polyester with PU backing, it offers superior abrasion, puncture, rip, and snag resistance, while being waterproof and windproof.
- Detachable hood with safety tear-away design
- Warm fleece collar
- Heavy-duty zipper with storm flap
- Dual reflective chest straps
- Double-stitched seams for durability
- Front and back vents for breathability
- Two zippered outside pockets and one inner chest pocket
- Adjustable wrist with hook-and-loop fastener
- Reflective material of 2 in in 4 in contrasting tape
- Compliant with CSA Z96-22 Class 1, Level 2 and ANSI/ISEA 107-2020 Type O, Class 1, ASTM D6413 (FR)
- Flame resistance tested per CAN/CGSB-4.2 No.27.10 & ASTM D6413