Learn More
MSC Viking Rain Jacket, X-Large, Black, Polyester, Flame-Resistant, 1 ea
Catalog No. 5000787287
Encompass
Viking Professional rain jacket is engineered for cold weather and provides flame and wind resistance. Constructed from polyester, this X-Large, black jacket offers reliable protection in demanding environments.
  • Features a detachable hood
  • Equipped with a zipper closure
  • Constructed with double-stitched seams for durability
  • Includes 2 functional pockets
  • Insulated for enhanced warmth in cold weather
  • Certified to ASTM F2302-08 standard
  • Complies with CSA Z96-09 Class 1, Level FR certification
Catalog No. 50-007-87287 Supplier MSC Supplier No. 42327494
Viking Professional rain jacket is engineered for cold weather and provides flame and wind resistance. Constructed from polyester, this X-Large, black jacket offers reliable protection in demanding environments.
  • Features a detachable hood
  • Equipped with a zipper closure
  • Constructed with double-stitched seams for durability
  • Includes 2 functional pockets
  • Insulated for enhanced warmth in cold weather
  • Certified to ASTM F2302-08 standard
  • Complies with CSA Z96-09 Class 1, Level FR certification