Learn More
Aviva Systems Biology Corp VIL1 Antibody - C-terminal region (ARP65001_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65001_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VIL1. It was validated on Western Blot. . Alternate names: D2S1471, VILApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-411