Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vilagletistat | 1542132-88-6 | 99.8% | 528.60 | C26H36N6O6 | 10 MG
SDP

Catalog No. 5000297914
Encompass_Preferred

Vilagletistat (ZED-1227) is a small-molecule, orally active transglutaminase 2 (TG2) inhibitor supplied for research use. It is provided as a high-purity powder in multiple package sizes and includes storage and stability guidance for powder and solutions.

  • High purity: reported 99.82% typical; example batch 99.97%.
  • CAS number 1542132-88-6.
  • Molecular formula C26H36N6O6 and molecular weight 528.60 g·mol^-1.
  • Available package sizes from 1 mg to 100 mg; larger quantities available by quote.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-97914 Supplier Medchemexpress LLC Supplier No. HY1935910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vilagletistat (ZED-1227) is a small-molecule, orally active transglutaminase 2 (TG2) inhibitor supplied for research use. It is provided as a high-purity powder in multiple package sizes and includes storage and stability guidance for powder and solutions.

  • High purity: reported 99.82% typical; example batch 99.97%.
  • CAS number 1542132-88-6.
  • Molecular formula C26H36N6O6 and molecular weight 528.60 g·mol^-1.
  • Available package sizes from 1 mg to 100 mg; larger quantities available by quote.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.