Learn More
Medchemexpress LLC Vilanterol trifenatate | 503070-58-4 | MFCD 21362970 | 99.3% | 774.77 g/mol | C44H49Cl2NO7 | 5 MG
SDP
Catalog No. 502030525
Encompass_Preferred
Vilanterol trifenatate is a long-acting β2-adrenergic receptor agonist supplied as a research reagent. It produces sustained activation of airway β2 receptors to increase cAMP and relax bronchial smooth muscle, and is provided as a high-purity solid for preclinical and pharmacological studies.
  • Long-acting β2-adrenergic receptor agonist suitable for airway pharmacology studies.
  • High reported purity (LCMS 99.3%).
  • White to off-white solid, convenient for formulation and analytical work.
  • Useful as a reference compound in receptor pharmacology and preclinical respiratory research.
  • For research use only; not for human or clinical applications.
Catalog No. 50-203-0525 Supplier Medchemexpress LLC Supplier No. HY14300A5MG
Vilanterol trifenatate is a long-acting β2-adrenergic receptor agonist supplied as a research reagent. It produces sustained activation of airway β2 receptors to increase cAMP and relax bronchial smooth muscle, and is provided as a high-purity solid for preclinical and pharmacological studies.
  • Long-acting β2-adrenergic receptor agonist suitable for airway pharmacology studies.
  • High reported purity (LCMS 99.3%).
  • White to off-white solid, convenient for formulation and analytical work.
  • Useful as a reference compound in receptor pharmacology and preclinical respiratory research.
  • For research use only; not for human or clinical applications.