Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vilazodone-d8 | 1794789-93-7 | 99.0% | C26H19D8N5O2 | 1 MG
SDP

Catalog No. 5000353540
Encompass_Preferred

Vilazodone-d8 is a deuterium-labeled vilazodone, which acts as a combined serotonin specific reuptake inhibitor (SSRI) and 5-HT1A receptor partial agonist. It can be used as a tracer and internal standard for quantitative analysis.

  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Targets 5-HT1A Receptor
  • Appears as a solid
  • Color: white to off-white
  • Soluble in H2O: ≥ 0.32 mg/mL (0.71 mM)

Catalog No. 50-003-53540 Supplier Medchemexpress LLC Supplier No. HY14261S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vilazodone-d8 is a deuterium-labeled vilazodone, which acts as a combined serotonin specific reuptake inhibitor (SSRI) and 5-HT1A receptor partial agonist. It can be used as a tracer and internal standard for quantitative analysis.

  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Targets 5-HT1A Receptor
  • Appears as a solid
  • Color: white to off-white
  • Soluble in H2O: ≥ 0.32 mg/mL (0.71 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.