Learn More
Medchemexpress LLC Vilazodone Hydrochloride | 163521-08-2 | 100.7% | C26H28ClN5O2 | 1 G
SDP
Catalog No. 5000355633
Encompass_Preferred
Vilazodone Hydrochloride is a serotonin transporter (SER) inhibitor and 5-HT1A receptor partial agonist. It is used to attenuate stress-induced potentiated startle and is also an option for the treatment of Major Depressive Disorder (MDD).
  • Serotonin transporter (SER) inhibitor
  • 5-HT1A receptor partial agonist
  • Attenuates stress-induced potentiated startle
  • Option for the treatment of major depressive disorder
  • Investigated in multiple clinical trials
Catalog No. 50-003-55633 Supplier Medchemexpress LLC Supplier No. HY142611G
Add to Cart
Add to Cart
Vilazodone Hydrochloride is a serotonin transporter (SER) inhibitor and 5-HT1A receptor partial agonist. It is used to attenuate stress-induced potentiated startle and is also an option for the treatment of Major Depressive Disorder (MDD).
  • Serotonin transporter (SER) inhibitor
  • 5-HT1A receptor partial agonist
  • Attenuates stress-induced potentiated startle
  • Option for the treatment of major depressive disorder
  • Investigated in multiple clinical trials