Learn More
Medchemexpress LLC Vilazodone Hydrochloride | 163521-08-2 | 100.7% | C26H28ClN5O2 | 100 MG
SDP
Catalog No. 5000355585
Encompass_Preferred
Vilazodone Hydrochloride is a serotonin transporter (SER) inhibitor and 5-HT1A receptor partial agonist. It is intended for research use only and not for patient use. In animal studies, it produced a characteristic behavioral syndrome, inhibited stress-induced vocalizations, and attenuated stress-induced potentiated startle.
  • Serotonin transporter (SER) inhibitor
  • 5-HT1A receptor partial agonist
  • Suitable for research use
  • Attenuates stress-induced potentiated startle in animal studies
  • Option for the treatment of Major Depressive Disorder (MDD)
Catalog No. 50-003-55585 Supplier Medchemexpress LLC Supplier No. HY14261100MG
Vilazodone Hydrochloride is a serotonin transporter (SER) inhibitor and 5-HT1A receptor partial agonist. It is intended for research use only and not for patient use. In animal studies, it produced a characteristic behavioral syndrome, inhibited stress-induced vocalizations, and attenuated stress-induced potentiated startle.
  • Serotonin transporter (SER) inhibitor
  • 5-HT1A receptor partial agonist
  • Suitable for research use
  • Attenuates stress-induced potentiated startle in animal studies
  • Option for the treatment of Major Depressive Disorder (MDD)