Learn More
Medchemexpress LLC Vilazodone hydrochloride | 163521-08-2 | 100.7% | C26H28ClN5O2 | 200 MG
SDP
Catalog No. 5000355634
Encompass_Preferred
Vilazodone hydrochloride is a serotonin transporter (SER) inhibitor and 5-HT1A receptor partial agonist. This product is for research use only.
  • Serotonin transporter (SER) inhibitor
  • 5-HT1A receptor partial agonist
  • Ships at room temperature
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture
Catalog No. 50-003-55634 Supplier Medchemexpress LLC Supplier No. HY14261200MG
Vilazodone hydrochloride is a serotonin transporter (SER) inhibitor and 5-HT1A receptor partial agonist. This product is for research use only.
  • Serotonin transporter (SER) inhibitor
  • 5-HT1A receptor partial agonist
  • Ships at room temperature
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture