Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vilazodone Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. V0149200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0149200MG 200 mg
V014950MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0149200MG Supplier TCI America Supplier No. V0149200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 163521-08-2
Molecular Formula C26H28ClN5O2
Molecular Weight (g/mol) 477.993
MDL Number MFCD00940014
InChI Key RPZBRGFNBNQSOP-UHFFFAOYSA-N
Synonym 5-[4-[4-(5-Cyano-1H-indol-3-yl)butyl]piperazin-1-yl]benzofuran-2-carboxamide Hydrochloride
PubChem CID 6918313
ChEBI CHEBI:70705
IUPAC Name 5-[4-[4-(5-cyano-1H-indol-3-yl)butyl]piperazin-1-yl]-1-benzofuran-2-carboxamide;hydrochloride
SMILES C1CN(CCN1CCCCC2=CNC3=C2C=C(C=C3)C#N)C4=CC5=C(C=C4)OC(=C5)C(=O)N.Cl

Specifications

Melting Point 279°C
Color White-Yellow
Quantity 200 mg
Formula Weight 477.99
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Vilazodone Hydrochloride
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.