Learn More
Cayman Chemical Vilazodone (hydrochloride), Purity: ≥95%, CAS: 163521-08-2, 100 mg
SDP
Catalog No. 502964075
Encompass_Preferred
An SSRI and a partial agonist of 5-HT1A (IC50 s = 0.2 and 0.5 nM, respectively); increases extracellular 5-HT in the rat ventral hippocampus and frontal cortex at 1 and 3 mg/kg, i.p.; decreases immobility in the forced swim test in both rats and mice at 1 mg/kg
Catalog No. 50-296-4075 Supplier Cayman Chemical Supplier No. 21547100 MG
An SSRI and a partial agonist of 5-HT1A (IC50 s = 0.2 and 0.5 nM, respectively); increases extracellular 5-HT in the rat ventral hippocampus and frontal cortex at 1 and 3 mg/kg, i.p.; decreases immobility in the forced swim test in both rats and mice at 1 mg/kg