Learn More
Cayman Chemical VIldaglIptIn carboxylIc ACD
SDP
Catalog No. 502987091
Encompass_Preferred
A major metabolite of vildagliptin; IC50 = 477 µM for DPP-4 in human Caco-2 cells; formed from hydrolysis of the cyano group on vildagliptin
Catalog No. 50-298-7091 Supplier Cayman Chemical Supplier No. 22088500 UG
A major metabolite of vildagliptin; IC50 = 477 µM for DPP-4 in human Caco-2 cells; formed from hydrolysis of the cyano group on vildagliptin