Learn More
Medchemexpress LLC Vildagliptin-d3 | 1217546-82-1 | C17H22D3N3O2 | 500 UG
SDP
Supplier:  Medchemexpress LLC HY14291S500UG
Encompass_Preferred
Vildagliptin-d3 is a deuterium labeled Vildagliptin. It is a potent, stable, selective dipeptidyl peptidase IV (DPP-IV) inhibitor with an IC50 of 3.5 nM in human Caco-2 cells. It possesses excellent oral bioavailability and potent antihyperglycemic activity. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-55510