Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Ideal for hospital and nursing home facilities
- Cotton-free design provides superior strength and absorbency
- Looped-end construction allows for maxium floor coverage while eliminating tangling, fraying, and linting
- Can be laundered for re-use
Specifications
Specifications
| Product Type | Looped End String Mop |
| Color | Blue |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Size | Medium |
| Description | 6 Row Stitched with 5 Ply Yarn |
| For Use With (Application) | General Use |
| Material | Synthetic Fiber |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?