Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ NanoTech™ micro Microfiber Cloths

Catalog No. 19032251 Shop All Vileda Professional Products
Change view
Click to view available options
Color:
Blue
Green
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-032-251 Blue
19-032-252 Green
19-032-253 Red
19-032-250 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-032-251 Supplier Vileda Professional™ Supplier No. 128601

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Superior cloths for superior wiping performance. Whichever environment you work in, you can be certain that microfiber wiping will improve your work efforts. Benefits are maximum performance, low cost-in-use and maximum hygiene as color coding prevents cross-contamination. NanoTech™ cloth is the solution for all environments where hygiene is critical.

  • Nanosilver particles embedded in endless microfibers
  • Proven cleaning efficacy
  • No growth of bacteria odors or germs on cloths for more than 24 hours
  • Extremely low linting
  • Omit tumble drying as cloths can be stored in moist condition up to 24 hours without bacteria odor growth
  • Keeps antibacterial properties after 300+ washing cycles
  • Color coded to prevent cross contamination

Specifications

Product Type Microfiber Cloth
Material Microfiber
Certifications/Compliance BMA
Color Blue
Dimensions (L x W) 38 x 40 cm (15 x 16 in.)
For Use With (Application) Hospitals, nursing homes, doctors surgeries, hotels, restaurants, public areas, schools and nurseries
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.