Learn More
Description
- High chemical resistance against chlorine, bleach, disinfectants, peroxides, etc.
- Non-woven structure
- Easy to rinse clean, hairs and dirt do not cling to surface
- Very low-linting fabric due to patented endless fiber technology
- Launderable up to 300+ wash cycles
Specifications
Specifications
| Color | Red |
| Dimensions (L x W) | 16 x 16 in. |
| For Use With (Application) | ISO Class 7, LAR, Production, Labs |
| Packaging Quantity | 5 Cloths /Pk., 20 Pk./Cs. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.