Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Freudenberg™ Vileda™ QuickStar micro Microfiber Cloths

Catalog No. 19000668 Shop All Vileda Professional Products
Change view
Click to view available options
Color:
Blue
Green
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-000-668 Yellow
19-000-665 Red
19-000-666 Blue
19-000-667 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-000-668 Supplier Freudenberg™ Supplier No. 100256

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for pre-preparation method of cloths in boxes/buckets with cleaning solution. Vileda Professional QuickStar micro Microfiber Cloths offers high chemical resistance against chlorine, bleach, disinfectants, peroxides.

  • High chemical resistance against chlorine, bleach, disinfectants, peroxides, etc.
  • Non-woven structure
  • Easy to rinse clean, hairs and dirt do not cling to surface
  • Very low-linting fabric due to patented endless fiber technology
  • Launderable up to 300+ wash cycles

Specifications

Color Yellow
Dimensions (L x W) 16 x 16 in.
For Use With (Application) ISO Class 7, LAR, Production, Labs
Packaging Quantity 5 Cloths /Pk., 20 Pk./Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.