Learn More
Aviva Systems Biology Corp VILL Antibody - N-terminal region (ARP63465_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63465_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VILL. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-287-976