Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Villella’s Reagent for Metal Etching Meets ASTM E407 Specification Etchant 80, Reagents

Catalog No. 01333399
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-399 500 mL
01-333-400 250 mL
01-333-402 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-333-399 Supplier Reagents Holdings Llc Supplier No. CV200200500C

missing translation for 'validMainframeMsg'

Villella’s Reagent for Metal Etching Meets ASTM E407 Specification Etchant 80, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 7732-18-5 , 67-63-0
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Vilellas
IUPAC Name ethanol
SMILES CCO

Specifications

Color Yellow
Percent Purity 1%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.8276
pH <1
Quantity 500 mL
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Villella’s Reagent for Metal Etching Meets ASTM E407 Specification Etchant 80
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.