Learn More
Villella’s Reagent for Metal Etching Meets ASTM E407 Specification Etchant 80, Reagents
Villella’s Reagent for Metal Etching Meets ASTM E407 Specification Etchant 80, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$114.00
Chemical Identifiers
| CAS | 64-17-5 , 7732-18-5 , 67-63-0 |
|---|---|
| Molecular Formula | C2H6O |
| Molecular Weight (g/mol) | 46.07 |
| InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
| Synonym | Vilellas |
| IUPAC Name | ethanol |
| SMILES | CCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-333-400
|
Reagents Holdings Llc
CV200200250C |
250 mL |
Each for $114.00
|
|
|||||
|
01-333-402
|
Reagents Holdings Llc
CV2002001C |
1 L | N/A | N/A | N/A | ||||
|
01-333-399
|
Reagents Holdings Llc
CV200200500C |
500 mL | N/A | N/A | N/A | ||||
Chemical Identifiers
| C2H6O | |
| LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
| ethanol |
Specifications
| Yellow | |
| C2H6O | |
| LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
| ethanol | |
| 1% | |
| Glass Bottle | |
| <1 | |
| Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.