Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Villella’s Reagent for Metal Etching Meets ASTM E407 Specification Etchant 80, Reagents

Villella’s Reagent for Metal Etching Meets ASTM E407 Specification Etchant 80, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc CV200200250C

Quantity 250 mL

 View more versions of this product

Catalog No. 01-333-400


 Restricted Item
Add to Cart

Description

Description

Specifications

Chemical Identifiers

64-17-5 , 7732-18-5 , 67-63-0
46.07
Vilellas
CCO
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol

Specifications

Yellow
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
1%
Glass Bottle
<1
Non-sterile
Villella’s Reagent for Metal Etching Meets ASTM E407 Specification Etchant 80
64-17-5 , 7732-18-5 , 67-63-0
Vilellas
CCO
46.07
Reagent
0.8276
250 mL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.