Learn More
Diagnostic Biosystems VILLIN Villin (1D2C3)
Catalog No. 502407329
Encompass_Preferred
Villin is One of the gelsolin family of calcium regulated actin binding proteins. Normally expressed in the brush border of epithelial cells lining the gastrointestinal tract, hepatobiliary tract and renal proximal convoluted tubules. In colorectal carcinoma, villin is highly expressed. In practice, villin can be included in the panel used for metastatic carcinoma to detect colorectal origin.
Catalog No. 50-240-7329 Supplier Diagnostic Biosystems Supplier No. PDM590
Villin is One of the gelsolin family of calcium regulated actin binding proteins. Normally expressed in the brush border of epithelial cells lining the gastrointestinal tract, hepatobiliary tract and renal proximal convoluted tubules. In colorectal carcinoma, villin is highly expressed. In practice, villin can be included in the panel used for metastatic carcinoma to detect colorectal origin.