Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7429MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: WB. Conjugate/Tag: Unconjugated. Clone: VIL1/1314. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: A549, HepG2 and HCT116 cells. Colon or Rectum. Cellular Staining: Cytoplasmic ;Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human Villin fragment of 133 amino acid residues (aa179-311). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 93kDa.Chromosome: 2q35. Gene Symbol: VIL1. Gene ID: 7429. Ensemble: ENSG00000127831. Uniprot: NP_009058.
Catalog No. 50-188-5270