Learn More
STEMCELL Technologies Anti-Human Villin 1 Antibody, Polyclonal, Size: 100 ul
SDP
Supplier:  STEMCELL Technologies 1001066
Encompass
This rabbit polyclonal antibody reacts with human villin 1 a calcium-regulated protein expressed in epithelial cells. Villin 1 is organized into six conserved domains which make up the villin core and a C-terminal headpiece domain which contains an F-actin binding site. Use this polyclonal antibody in applications requiring detection of villin 1 such as western blotting (WB) assays and immunohistochemistry (IHC). It is verified for use with immunocytochemistry (ICC).
Catalog No. 50-239-0220