Learn More
ABclonal Technology Villin1 Rabbit mAb
SDP
Catalog No. 5000007778
Encompass_Preferred
This gene encodes a member of a family of calcium-regulated actin-binding proteins This protein represents a dominant part of the brush border cytoskeleton which functions in the capping severing and bundling of actin filaments Two mRNAs of 2 7 kb and 3 5 kb have been observed they result from utilization of alternate poly-adenylation signals present in the terminal exon
Catalog No. 50-000-07778 Supplier ABclonal Technology Supplier No. A23273100UL
This gene encodes a member of a family of calcium-regulated actin-binding proteins This protein represents a dominant part of the brush border cytoskeleton which functions in the capping severing and bundling of actin filaments Two mRNAs of 2 7 kb and 3 5 kb have been observed they result from utilization of alternate poly-adenylation signals present in the terminal exon