Learn More
Cayman Chemical Vilanterol (triphenylacetate), Purity: ≥95%, CAS: 503070-58-4, 50 mg
SDP
Catalog No. 502917329
Encompass_Preferred
A β2-AR agonist; selectively induces cAMP accumulation in CHO cells expressing the β2-AR over the β1- and β3-ARs (EC50s = 0.4, 398, and 794 nM for the human receptors, respectively); inhibits contractions induced by electrical stimulation in isolated superfused guinea pig trachea strips; inhibits histamine-induced bronchospasms in guinea pigs when administered via nebulization (EC50 = 30 µM)
Catalog No. 50-291-7329 Supplier Cayman Chemical Supplier No. 3953650 MG
A β2-AR agonist; selectively induces cAMP accumulation in CHO cells expressing the β2-AR over the β1- and β3-ARs (EC50s = 0.4, 398, and 794 nM for the human receptors, respectively); inhibits contractions induced by electrical stimulation in isolated superfused guinea pig trachea strips; inhibits histamine-induced bronchospasms in guinea pigs when administered via nebulization (EC50 = 30 µM)