Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Viloxazine hydrochloride | 35604-67-2 | MFCD00661100 | 98.9% | 273.75 g/mol | C13H20ClNO3 | 25MG
SDP

Catalog No. 5000239099
Encompass_Preferred

Viloxazine hydrochloride is the hydrochloride salt of viloxazine, a selective norepinephrine reuptake inhibitor supplied as a research-grade solid reference compound. It is intended for biochemical and pharmacological studies and includes analytical and storage guidance. For research use only; not for human or veterinary use.

  • High purity: 98.9% (reported by supplier)
  • Molecular formula: C13H20ClNO3
  • Molecular weight: 273.75 g/mol
  • Form: solid powder suitable for formulation or assay
  • Storage: store sealed, away from moisture; recommended 4°C; in solvent, -80°C (6 months) or -20°C (1 month)
  • Available in small quantities suitable for research (for example, 25 mg)

Catalog No. 50-002-39099 Supplier Medchemexpress LLC Supplier No. HY12578425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Viloxazine hydrochloride is the hydrochloride salt of viloxazine, a selective norepinephrine reuptake inhibitor supplied as a research-grade solid reference compound. It is intended for biochemical and pharmacological studies and includes analytical and storage guidance. For research use only; not for human or veterinary use.

  • High purity: 98.9% (reported by supplier)
  • Molecular formula: C13H20ClNO3
  • Molecular weight: 273.75 g/mol
  • Form: solid powder suitable for formulation or assay
  • Storage: store sealed, away from moisture; recommended 4°C; in solvent, -80°C (6 months) or -20°C (1 month)
  • Available in small quantities suitable for research (for example, 25 mg)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.