Learn More
Medchemexpress LLC Viloxazine hydrochloride | 35604-67-2 | MFCD00661100 | 98.9% | 273.75 g/mol | C13H20ClNO3 | 50MG
SDP
Catalog No. 5000239112
Encompass_Preferred
Viloxazine hydrochloride is a research-grade small molecule used as a selective norepinephrine transporter (NET) inhibitor and a modulator of serotonin (5-HT) receptors. Presented as the hydrochloride salt (CAS 35604-67-2) and supplied for preclinical laboratory use, it is commonly employed as a tool compound in studies of norepinephrine and serotonin signaling and in ADHD-related research.
  • Research-grade hydrochloride salt with defined purity for laboratory use.
  • Selective norepinephrine transporter inhibitor activity (IC50 = 0.26 μM).
  • Modulates 5-HT receptors, including 5-HT2B antagonist and 5-HT2C agonist actions.
  • Available in small pack sizes suitable for assays, including a 50 mg pack.
  • Provided with molecular and analytical data to support traceability and reproducibility.
Catalog No. 50-002-39112 Supplier Medchemexpress LLC Supplier No. HY12578450MG
Viloxazine hydrochloride is a research-grade small molecule used as a selective norepinephrine transporter (NET) inhibitor and a modulator of serotonin (5-HT) receptors. Presented as the hydrochloride salt (CAS 35604-67-2) and supplied for preclinical laboratory use, it is commonly employed as a tool compound in studies of norepinephrine and serotonin signaling and in ADHD-related research.
  • Research-grade hydrochloride salt with defined purity for laboratory use.
  • Selective norepinephrine transporter inhibitor activity (IC50 = 0.26 μM).
  • Modulates 5-HT receptors, including 5-HT2B antagonist and 5-HT2C agonist actions.
  • Available in small pack sizes suitable for assays, including a 50 mg pack.
  • Provided with molecular and analytical data to support traceability and reproducibility.